Assay Details
Target Gene Details
Entrez Gene ID: | 67429 |
Gene Name: | NudC domain containing 1 |
Gene Aliases: |
4921532K09Rik, AA407246, AW260430, AW556235, CML66-L, Cml66 |
Location: |
Chr.15:44375227-44428307 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudcd1 | NM_001113554.1 | 10 | 2239 | NP_001107026.1 |
| NM_026149.4 | 10 | 2242 | NP_080425.3 | |
| AF521132.1 | 10 | 2240 | AAQ08822.1 | |
| AK014996.2 | 10 | 2242 | ||
| AK077673.1 | 10 | 1824 | BAC36947.1 | |
| AK167855.1 | 10 | 1774 | BAE39873.1 | |
| AK168122.1 | 10 | 1774 | BAE40093.1 | |
| BC031583.1 | 10 | 1808 | AAH31583.1 |

Genomic Map