This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00628419_cn
See other GATA1 CNV Assays ›
Gene Symbol
Gata1
Assay Reference Genome
Location

Chr.X:7959972 on build GRCm38
Cytoband
XA1.1
Assay ID Mm00628419_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

14460

Gene Name:

GATA binding protein 1

Gene Aliases:

Gata-1, Gf-1, eryf1

Location:

Chr.X:7959260-7976677 on Build GRCm38

Assay Gene Location:

Within Exon 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
Gata1 NM_008089.2 6 1190 NP_032115.1
AK146915.1 6 1175 BAE27527.1
BC052653.1 6 1157 AAH52653.1
X15763.1 6 1095 CAA33769.1

Back To Top

More Information


Set Membership:

Intragenic Exonic

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
cell-cell signaling
negative regulation of cell proliferation
male gonad development
regulation of glycoprotein biosynthetic process
regulation of definitive erythrocyte differentiation
myeloid cell differentiation
erythrocyte differentiation
megakaryocyte differentiation
platelet formation
basophil differentiation
eosinophil differentiation
negative regulation of bone mineralization
positive regulation of osteoblast proliferation
embryonic hemopoiesis
eosinophil fate commitment
negative regulation of apoptotic process
positive regulation of erythrocyte differentiation
positive regulation of transcription from RNA polymerase II promoter
cell development
erythrocyte development
positive regulation of peptidyl-tyrosine phosphorylation
platelet aggregation
transcriptional activation by promoter-enhancer looping
dendritic cell differentiation
cellular response to thyroid hormone stimulus
negative regulation of transcription regulatory region DNA binding
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
transcription regulatory region sequence-specific DNA binding
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
enhancer sequence-specific DNA binding
p53 binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
DNA binding, bending
chromatin DNA binding
sequence-specific DNA binding
metal ion binding
C2H2 zinc finger domain binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-vvsgg:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0