Assay Details
Target Gene Details
Entrez Gene ID: | 75656 |
Gene Name: | RIKEN cDNA 1700020A23 gene |
Gene Aliases: |
- |
Location: |
Chr.2:130405259-130406074 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700020A23Rik | NM_001163483.1 | NP_001156955.1 | ||
| NM_029375.1 | NP_083651.1 |

Genomic Map