Assay Details
Target Gene Details
Entrez Gene ID: | 77994 |
Gene Name: | Mir99a and Mirlet7c-1 host gene (non-protein coding) |
Gene Aliases: |
2810055G20Rik, D730043B02Rik |
Location: |
Chr.16:77329328-77558428 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir99ahg | NR_015543.2 | |||
| AK083771.1 |

Genomic Map