Assay Details
Target Gene Details
Entrez Gene ID: | 5184 |
Gene Name: | peptidase D |
Gene Aliases: |
PROLIDASE |
Location: |
Chr.19:33386949-33521893 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PEPD | NM_000285.3 | 15 | 1572 | NP_000276.2 |
| NM_001166056.1 | 13 | 1449 | NP_001159528.1 | |
| NM_001166057.1 | 13 | 1380 | NP_001159529.1 | |
| AK057538.1 | 2 | 1306 | ||
| AK225037.1 | 15 | 1453 | ||
| AK290756.1 | 15 | 1453 | ||
| AK290781.1 | 15 | 1468 | ||
| AK291561.1 | 15 | 1470 | ||
| AK293126.1 | 13 | 1275 | ||
| AK294619.1 | 13 | 1347 | ||
| AW512991.1 | 1 | 448 | ||
| BC004305.1 | 15 | 1443 | AAH04305.1 | |
| BC015027.1 | 15 | 1473 | AAH15027.1 | |
| BC028295.1 | 15 | 1444 | AAH28295.1 | |
| BT006692.1 | AAP35338.1 | |||
| CR541669.1 | CAG46470.1 | |||
| J04605.1 | 15 | 1455 | AAA60064.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758755 | Chr.19:33286168 - 34090656 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map