Assay Details
Target Gene Details
Entrez Gene ID: | 165829 |
Gene Name: | G protein-coupled receptor 156 |
Gene Aliases: |
GABABL, PGR28 |
Location: |
Chr.3:120165481-120285122 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR156 | NM_001168271.1 | 9 | 2471 | NP_001161743.1 |
| NM_153002.2 | 9 | 2483 | NP_694547.2 | |
| XM_005247165.2 | 10 | 2760 | XP_005247222.1 | |
| XM_011512484.2 | 11 | 2833 | XP_011510786.1 | |
| XM_011512487.1 | 9 | 2385 | XP_011510789.1 | |
| XM_011512488.2 | 2 | 1452 | XP_011510790.1 | |
| XM_017005795.1 | 10 | 2796 | XP_016861284.1 | |
| XM_017005796.1 | 10 | 2805 | XP_016861285.1 | |
| XM_017005797.1 | 10 | 2748 | XP_016861286.1 | |
| AB209362.1 | 4 | 1647 | BAD92599.1 | |
| AF488739.1 | 9 | 2286 | AAN03795.1 | |
| BC113701.1 | 9 | 2306 | ||
| BC143606.1 | 9 | 2294 | ||
| BX115336.1 | 1 | 232 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013788 | Chr.3:120147875 - 120193450 on Build GRCh38 | Gain |
|
| nsv829697 | Chr.3:120084009 - 120199588 on Build GRCh38 | Loss |
|
| nsv3967 | Chr.3:120138861 - 120184268 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map