Assay Details
Target Gene Details
Entrez Gene ID: | 89910 |
Gene Name: | ubiquitin protein ligase E3B |
Gene Aliases: |
BPIDS, KOS |
Location: |
Chr.12:109477410-109543628 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE3B | NM_130466.3 | NP_569733.2 | ||
| NM_183415.2 | NP_904324.1 | |||
| XM_005253987.2 | XP_005254044.1 | |||
| XM_006719681.3 | XP_006719744.1 | |||
| XM_006719682.2 | XP_006719745.1 | |||
| XM_011538959.2 | XP_011537261.1 | |||
| XM_011538961.1 | XP_011537263.1 | |||
| XM_017020195.1 | XP_016875684.1 | |||
| AF130072.1 | ||||
| AF251046.2 | AAK28419.2 | |||
| AK307385.1 | ||||
| AL096740.1 | CAH56410.1 | |||
| BC032301.1 | ||||
| BC141880.1 | ||||
| BC150637.1 | ||||
| BX537403.1 | CAD97645.1 | |||
| DQ570810.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3630719 | Chr.12:109485009 - 109700854 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map