Assay Details
Target Gene Details
Entrez Gene ID: | 64784 |
Gene Name: | CREB regulated transcription coactivator 3 |
Gene Aliases: |
TORC-3, TORC3 |
Location: |
Chr.15:90529886-90645345 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRTC3 | NM_001042574.2 | 10 | 1032 | NP_001036039.1 |
| NM_022769.4 | 10 | 1032 | NP_073606.3 | |
| XM_005254968.2 | 10 | 1000 | XP_005255025.1 | |
| XM_011521906.2 | 9 | 904 | XP_011520208.1 | |
| AK025521.1 | 3 | 199 | BAB15160.1 | |
| AK074170.1 | 7 | 503 | BAB84996.1 | |
| AK090443.1 | 10 | 992 | BAC03424.1 | |
| AK225300.1 | 3 | 199 | ||
| AY360173.1 | 10 | 853 | AAQ98858.1 | |
| BC074730.2 | AAH74730.2 | |||
| BC074731.2 | ||||
| BP238198.1 | 3 | 200 | ||
| DB288138.1 | 1 | 38 |
Target Gene Details
Entrez Gene ID: | 101926895 |
Gene Name: | CRTC3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:90620009-90717140 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRTC3-AS1 | NR_120372.1 | |||
| BG472948.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045042 | Chr.15:90453811 - 90841192 on Build GRCh38 | Gain |
|
| nsv509582 | Chr.15:90578822 - 90663170 on Build GRCh38 | Insertion |
|
| nsv833088 | Chr.15:90469969 - 90645804 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map