Assay Details
Target Gene Details
Entrez Gene ID: | 9650 |
Gene Name: | mitochondrial fission regulator 1 |
Gene Aliases: |
CHPPR, FAM54A2 |
Location: |
Chr.8:65643873-65771252 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTFR1 | XM_006716484.2 | XP_006716547.2 | ||
| XM_011517626.1 | XP_011515928.1 | |||
| XM_011517627.2 | XP_011515929.1 | |||
| XM_011517628.2 | XP_011515930.1 |
Target Gene Details
Entrez Gene ID: | 5150 |
Gene Name: | phosphodiesterase 7A |
Gene Aliases: |
HCP1, PDE7 |
Location: |
Chr.8:65714334-65841734 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE7A | NM_001242318.2 | NP_001229247.1 | ||
| NM_002603.3 | NP_002594.1 | |||
| XM_011517540.2 | XP_011515842.1 | |||
| XM_017013538.1 | XP_016869027.1 | |||
| AF332652.1 | AAK57640.1 | |||
| AK290801.1 | ||||
| AK292680.1 | ||||
| BC126360.1 | ||||
| U67932.1 | AAB65772.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv395873 | Chr.8:65723621 - 65724253 on Build GRCh38 | Deletion |
|
| dgv247n6 | Chr.8:65724352 - 65724778 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map