Assay Details
Target Gene Details
Entrez Gene ID: | 80342 |
Gene Name: | TRAF3 interacting protein 3 |
Gene Aliases: |
T3JAM |
Location: |
Chr.1:209756032-209782323 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAF3IP3 | NM_001320144.1 | NP_001307073.1 | ||
| NM_025228.3 | NP_079504.2 | |||
| NR_109871.1 | ||||
| XM_011510018.2 | XP_011508320.1 | |||
| XM_011510019.1 | XP_011508321.1 | |||
| XM_017002399.1 | XP_016857888.1 | |||
| XM_017002400.1 | XP_016857889.1 | |||
| AK122792.1 | ||||
| AK225762.1 | ||||
| AK300555.1 | ||||
| AK312755.1 | ||||
| AL049667.1 | CAB41242.1 | |||
| AL558384.3 | ||||
| BC110302.1 | AAI10303.1 | |||
| BX648450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006632 | Chr.1:209712857 - 209797010 on Build GRCh38 | Gain |
|
| nsv1001588 | Chr.1:209735531 - 209764898 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map