Assay Details
Target Gene Details
Entrez Gene ID: | 2113 |
Gene Name: | ETS proto-oncogene 1, transcription factor |
Gene Aliases: |
ETS-1, EWSR2, c-ets-1, p54 |
Location: |
Chr.11:128458761-128587593 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ETS1 | NM_001143820.1 | 10 | 2536 | NP_001137292.1 |
| NM_001162422.1 | 4 | 1987 | NP_001155894.1 | |
| NM_005238.3 | 8 | 2635 | NP_005229.1 | |
| XM_011542649.2 | 11 | 2672 | XP_011540951.1 | |
| XM_011542650.1 | 9 | 3127 | XP_011540952.1 | |
| XM_011542651.1 | 9 | 2306 | XP_011540953.1 | |
| XM_011542652.1 | 7 | 2764 | XP_011540954.1 | |
| XM_017017314.1 | 10 | 2525 | XP_016872803.1 | |
| XM_017017315.1 | 9 | 2451 | XP_016872804.1 | |
| XM_017017316.1 | 9 | 18642 | XP_016872805.1 | |
| XM_017017317.1 | 9 | 2310 | XP_016872806.1 | |
| AK001630.1 | 1 | 232 | ||
| AL832693.1 | 1 | 304 | ||
| BX640634.1 | 10 | 2536 | CAE45783.1 |
More Information
Set Membership: |
|

Genomic Map