Assay Details
Target Gene Details
Entrez Gene ID: | 84268 |
Gene Name: | RPA interacting protein |
Gene Aliases: |
HRIP, RIP |
Location: |
Chr.17:5419185-5433020 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPAIN | NM_001033002.3 | 1 | 467 | NP_001028174.2 |
| NM_001160243.1 | 1 | 467 | NP_001153715.1 | |
| NM_001160244.1 | 1 | 467 | NP_001153716.1 | |
| NM_001160246.1 | 1 | 467 | NP_001153718.1 | |
| NM_001160266.1 | 1 | 467 | NP_001153738.1 | |
| NR_027679.1 | 1 | 467 | ||
| NR_027682.1 | 1 | 467 | ||
| NR_027683.1 | 1 | 467 | ||
| NR_027684.1 | 1 | 467 | ||
| NR_027685.1 | 1 | 467 | ||
| AK226044.1 | 1 | 223 | ||
| AW268632.1 | ||||
| AY775314.1 | 1 | 467 | AAX14368.1 | |
| AY775315.1 | 1 | 467 | AAX14369.1 | |
| AY775316.1 | 1 | 467 | AAX14370.1 | |
| AY775317.1 | 1 | 467 | AAX14371.1 | |
| AY775318.1 | 1 | 467 | AAX14372.1 | |
| AY775319.1 | 1 | 467 | AAX14373.1 | |
| AY775320.1 | 1 | 467 | AAX14374.1 | |
| AY775321.1 | 1 | 467 | AAX14375.1 | |
| AY775323.1 | 1 | 467 | AAX14377.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map