Assay Details
Target Gene Details
Entrez Gene ID: | 732 |
Gene Name: | complement component 8, beta polypeptide |
Gene Aliases: |
C82 |
Location: |
Chr.1:56929210-56974383 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8B | NM_000066.3 | 7 | 1206 | NP_000057.2 |
| NM_001278543.1 | 8 | 1428 | NP_001265472.1 | |
| NM_001278544.1 | 8 | 1335 | NP_001265473.1 | |
| XM_017002235.1 | 8 | 1274 | XP_016857724.1 | |
| AK292740.1 | 7 | 1061 | ||
| AK298425.1 | 8 | 1252 | ||
| AK298444.1 | 8 | 1428 | ||
| AK313382.1 | 7 | 1078 | ||
| AK316209.1 | 8 | 1425 | ||
| BC020701.1 | 7 | 1081 | ||
| BC130575.1 | 7 | 1039 | ||
| M16973.1 | 7 | 1041 | AAA51862.1 | |
| X04393.1 | 6 | 876 | CAA27981.1 |
More Information
Set Membership: |
|

Genomic Map