Assay Details
Target Gene Details
Entrez Gene ID: | 144108 |
Gene Name: | SPT2 chromatin protein domain containing 1 |
Gene Aliases: |
Spt2 |
Location: |
Chr.11:18606401-18634823 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPTY2D1 | NM_194285.2 | 6 | 4876 | NP_919261.2 |
| XM_011519919.2 | 6 | 4734 | XP_011518221.1 | |
| AK096694.1 | 2 | 2780 | ||
| AL834393.1 | 3 | 2926 | CAD39055.1 | |
| BC056261.1 | AAH56261.1 | |||
| BX538046.1 | 4 | 3255 | CAD97985.1 | |
| BX641102.1 | 6 | 4777 | CAE46047.1 | |
| BX647798.1 | 6 | 4876 | ||
| BX648114.1 | 4 | 4455 | CAH10772.1 | |
| BX648478.1 | 6 | 4798 | ||
| CR749626.1 | 4 | 4319 | CAH18420.1 |
Target Gene Details
Entrez Gene ID: | 100506540 |
Gene Name: | SPTY2D1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:18599804-18610255 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPTY2D1-AS1 | NR_038360.1 | |||
| AK097654.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553633 | Chr.11:18290154 - 19036904 on Build GRCh38 | Gain |
|
| nsv516413 | Chr.11:18606871 - 18622211 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map