Assay Details
Target Gene Details
Entrez Gene ID: | 134218 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C21 |
Gene Aliases: |
DNAJA5, GS3, JJJ1 |
Location: |
Chr.5:34929540-34958964 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC21 | NM_001012339.2 | NP_001012339.2 | ||
| NM_194283.3 | NP_919259.3 | |||
| XM_005248249.4 | XP_005248306.2 | |||
| XM_005248250.3 | XP_005248307.2 | |||
| XM_011513965.2 | XP_011512267.2 | |||
| XM_011513966.2 | XP_011512268.2 | |||
| AK127749.1 | BAC87112.1 | |||
| AY177688.1 | AAO46910.1 | |||
| AY258422.1 | AAP81807.1 | |||
| BC050472.1 | AAH50472.1 | |||
| BC107577.1 | AAI07578.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3604691 | Chr.5:34934667 - 35064045 on Build GRCh38 | Gain |
|
| nsv597809 | Chr.5:34853860 - 34957339 on Build GRCh38 | Gain |
|
| dgv5629n100 | Chr.5:34798496 - 34983041 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map