Assay Details
Target Gene Details
Entrez Gene ID: | 81033 |
Gene Name: | potassium voltage-gated channel subfamily H member 6 |
Gene Aliases: |
ERG-2, ERG2, HERG2, Kv11.2, hERG-2 |
Location: |
Chr.17:63523334-63548977 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNH6 | NM_001278919.1 | NP_001265848.1 | ||
| NM_001278920.1 | NP_001265849.1 | |||
| NM_030779.3 | NP_110406.1 | |||
| NM_173092.2 | NP_775115.1 | |||
| XM_011525308.2 | XP_011523610.1 | |||
| XM_011525309.2 | XP_011523611.1 | |||
| XM_011525310.2 | XP_011523612.1 | |||
| XM_011525311.1 | XP_011523613.1 | |||
| XM_011525312.1 | XP_011523614.1 | |||
| XM_011525313.1 | XP_011523615.1 | |||
| XM_017025175.1 | XP_016880664.1 | |||
| XM_017025176.1 | XP_016880665.1 | |||
| XM_017025177.1 | XP_016880666.1 | |||
| XM_017025178.1 | XP_016880667.1 | |||
| XM_017025179.1 | XP_016880668.1 | |||
| XM_017025180.1 | XP_016880669.1 | |||
| AF311913.1 | AAG40871.1 | |||
| AK091877.1 | BAC03764.1 | |||
| AK296498.1 | ||||
| AK298363.1 | ||||
| BC006334.2 | AAH06334.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3640998 | Chr.17:63526371 - 63539713 on Build GRCh38 | Loss |
|
| nsv952355 | Chr.17:63522640 - 63550539 on Build GRCh38 | Deletion |
|
| esv2675657 | Chr.17:63526487 - 63539627 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map