Assay Details
Target Gene Details
Entrez Gene ID: | 1386 |
Gene Name: | activating transcription factor 2 |
Gene Aliases: |
CRE-BP1, CREB-2, CREB2, HB16, TREB7 |
Location: |
Chr.2:175072250-175168206 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATF2 | NM_001256090.1 | 15 | 1996 | NP_001243019.1 |
| NM_001256091.1 | 14 | 1921 | NP_001243020.1 | |
| NM_001256092.1 | 13 | 1864 | NP_001243021.1 | |
| NM_001256093.1 | 13 | 1896 | NP_001243022.1 | |
| NM_001880.3 | 14 | 1961 | NP_001871.2 | |
| NR_045768.1 | 17 | 2158 | ||
| NR_045769.1 | 17 | 2190 | ||
| NR_045770.1 | 16 | 2043 | ||
| NR_045771.1 | 15 | 2035 | ||
| NR_045772.1 | 15 | 1973 | ||
| NR_045773.1 | 14 | 1938 | ||
| NR_045774.1 | 13 | 1846 | ||
| AI200584.1 | 1 | 194 | ||
| AK128731.1 | 14 | 1861 | ||
| AY029364.1 | AAK55760.1 | |||
| BC107698.1 | 13 | 1831 | AAI07699.1 | |
| BC130335.1 | ||||
| BC130337.1 | ||||
| BX648469.1 | 13 | 1751 | ||
| DQ003036.1 | AAY17202.1 | |||
| DQ003037.1 | AAY17203.1 | |||
| DQ003038.1 | AAY17204.1 | |||
| DQ003039.1 | AAY17205.1 | |||
| DQ003040.1 | AAY17206.1 | |||
| DQ003041.1 | AAY17207.1 | |||
| DQ003042.1 | AAY17208.1 | |||
| DQ003043.1 | AAY17209.1 | |||
| DQ003044.1 | AAY17210.1 | |||
| DQ003045.1 | AAY17211.1 | |||
| DQ003046.1 | AAY17212.1 | |||
| DQ003047.1 | AAY17213.1 | |||
| DQ003048.1 | AAY17214.1 | |||
| DQ003049.1 | AAY17215.1 | |||
| M31630.1 | 6 | 1032 | AAA35951.1 | |
| X15875.1 | CAA33886.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575313 | Chr.2:174591458 - 175085724 on Build GRCh38 | Gain |
|
| nsv1012709 | Chr.2:175023400 - 175840083 on Build GRCh38 | Gain |
|
| esv3390245 | Chr.2:175074493 - 175074507 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map