Assay Details
Target Gene Details
Entrez Gene ID: | 1240 |
Gene Name: | chemerin chemokine-like receptor 1 |
Gene Aliases: |
CHEMERINR, ChemR23, DEZ, RVER1 |
Location: |
Chr.12:108288044-108339348 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMKLR1 | NM_001142343.1 | 4 | 1922 | NP_001135815.1 |
| NM_001142344.1 | 3 | 1709 | NP_001135816.1 | |
| NM_001142345.1 | 3 | 1536 | NP_001135817.1 | |
| NM_004072.2 | 3 | 1846 | NP_004063.1 | |
| XM_017018820.1 | 2 | 1664 | XP_016874309.1 | |
| U79526.1 | AAC51258.1 | |||
| U79527.1 | AAC51259.1 |
More Information
Set Membership: |
|

Genomic Map