Assay Details
Target Gene Details
Entrez Gene ID: | 6000 |
Gene Name: | regulator of G-protein signaling 7 |
Gene Aliases: |
- |
Location: |
Chr.1:240775193-241357374 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468650 | Chr.1:240682824 - 240804127 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map