Assay Details
Target Gene Details
Entrez Gene ID: | 10486 |
Gene Name: | CAP, adenylate cyclase-associated protein, 2 (yeast) |
Gene Aliases: |
- |
Location: |
Chr.6:17393216-17557792 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAP2 | NM_006366.2 | NP_006357.1 | ||
| XM_011514233.2 | XP_011512535.1 | |||
| AK293134.1 | ||||
| AK294215.1 | ||||
| AK295600.1 | ||||
| AK297538.1 | ||||
| AK308461.1 | ||||
| AK312362.1 | ||||
| AL832154.1 | CAI46134.1 | |||
| BC008481.1 | AAH08481.1 | |||
| CR457022.1 | CAG33303.1 | |||
| U02390.1 | AAA20587.1 |
Target Gene Details
Entrez Gene ID: | 101928491 |
Gene Name: | uncharacterized LOC101928491 |
Gene Aliases: |
- |
Location: |
Chr.6:17501360-17511265 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928491 | NR_110855.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5922n100 | Chr.6:17490857 - 17921391 on Build GRCh38 | Gain |
|
| nsv516012 | Chr.6:17459945 - 17876993 on Build GRCh38 | Gain+Loss |
|
| nsv601046 | Chr.6:17492862 - 17893291 on Build GRCh38 | Gain |
|
| dgv1715e212 | Chr.6:17457368 - 17679729 on Build GRCh38 | Gain |
|
| nsv1027813 | Chr.6:17457369 - 17630059 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map