Assay Details
Target Gene Details
Entrez Gene ID: | 9227 |
Gene Name: | lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase) |
Gene Aliases: |
LCA14 |
Location: |
Chr.4:154740841-154753119 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRAT | NM_001301645.1 | 3 | 3747 | NP_001288574.1 |
| NM_004744.4 | 3 | 3941 | NP_004735.2 | |
| AY546085.1 | 2 | 4015 | AAS49412.1 | |
| AY546086.1 | 3 | 3912 | AAS49413.1 |
More Information
Set Membership: |
|

Genomic Map