Assay Details
Target Gene Details
Entrez Gene ID: | 11100 |
Gene Name: | heterogeneous nuclear ribonucleoprotein U like 1 |
Gene Aliases: |
E1B-AP5, E1BAP5, HNRPUL1 |
Location: |
Chr.19:41262476-41307783 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26941 | Chr.19:41264323 - 41309377 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map