Assay Details
Target Gene Details
Entrez Gene ID: | 4059 |
Gene Name: | basal cell adhesion molecule (Lutheran blood group) |
Gene Aliases: |
AU, CD239, LU, MSK19 |
Location: |
Chr.19:44809059-44821421 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCAM | NM_001013257.2 | 12 | 1633 | NP_001013275.1 |
| NM_005581.4 | 12 | 1633 | NP_005572.2 | |
| BC050450.1 | 12 | 1572 | AAH50450.1 | |
| HE572749.1 | 12 | 1567 | ||
| X80026.1 | 12 | 1573 | CAA56327.1 | |
| X83425.1 | 12 | 1589 | CAA58449.1 |
More Information
Set Membership: |
|

Genomic Map