Assay Details
Target Gene Details
Entrez Gene ID: | 91662 |
Gene Name: | NLR family pyrin domain containing 12 |
Gene Aliases: |
CLR19.3, FCAS2, NALP12, PAN6, PYPAF7, RNO, RNO2 |
Location: |
Chr.19:53793584-53824403 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRP12 | NM_001277126.1 | 3 | 1263 | NP_001264055.1 |
| NM_001277129.1 | 3 | 1263 | NP_001264058.1 | |
| NM_144687.3 | 3 | 1263 | NP_653288.1 | |
| XM_011527479.1 | 3 | 1263 | XP_011525781.1 | |
| XM_011527480.1 | 3 | 1263 | XP_011525782.1 | |
| XM_011527482.1 | 3 | 1263 | XP_011525784.1 | |
| XM_017027460.1 | 3 | 1263 | XP_016882949.1 | |
| XM_017027461.1 | 3 | 1263 | XP_016882950.1 | |
| XM_017027462.1 | 3 | 1263 | XP_016882951.1 | |
| XM_017027463.1 | 4 | 1036 | XP_016882952.1 | |
| XM_017027464.1 | 3 | 834 | XP_016882953.1 | |
| XM_017027465.1 | 4 | 1044 | XP_016882954.1 | |
| XM_017027466.1 | 4 | 1041 | XP_016882955.1 | |
| XM_017027467.1 | 3 | 836 | XP_016882956.1 | |
| AK095460.1 | 3 | 1178 | ||
| AK290772.1 | 3 | 1045 | ||
| AY095146.1 | 3 | 1210 | AAM18227.1 | |
| AY116204.1 | 3 | 1254 | AAM75142.1 | |
| AY116205.1 | 3 | 1254 | AAM75143.1 | |
| AY116206.1 | 3 | 1254 | AAM75144.1 | |
| AY116207.1 | 3 | 1254 | AAM75145.1 | |
| AY154467.1 | 3 | 1210 | AAO18163.1 | |
| BC028069.1 | 3 | 1153 | AAH28069.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064558 | Chr.19:53773468 - 53957491 on Build GRCh38 | Gain |
|
| esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map