Assay Details
Target Gene Details
Entrez Gene ID: | 7469 |
Gene Name: | negative elongation factor complex member A |
Gene Aliases: |
NELF-A, P/OKcl.15, WHSC2 |
Location: |
Chr.4:1982714-2009972 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NELFA | NM_005663.4 | NP_005654.3 | ||
| XM_017008589.1 | XP_016864078.1 | |||
| AF101434.1 | AAC72982.1 | |||
| AK001304.1 | ||||
| AK094040.1 | ||||
| AK126056.1 | ||||
| AK222853.1 | BAD96573.1 | |||
| BC002764.2 | AAH02764.2 | |||
| DB082393.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv822441 | Chr.4:1991240 - 2080440 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map