Assay Details
Target Gene Details
Entrez Gene ID: | 10550 |
Gene Name: | ADP ribosylation factor like GTPase 6 interacting protein 5 |
Gene Aliases: |
DERP11, GTRAP3-18, HSPC127, JWA, PRAF3, Yip6b, addicsin, hp22, jmx |
Location: |
Chr.3:69084939-69106088 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL6IP5 | NM_006407.3 | NP_006398.1 | ||
| AB014765.1 | BAC66462.1 | |||
| AB052638.1 | BAD83603.1 | |||
| AB097051.1 | BAC77404.1 | |||
| AF070523.1 | AAC64360.1 | |||
| AF115965.1 | AAP97237.1 | |||
| AF125530.1 | AAF17224.1 | |||
| AF161476.1 | AAF29091.1 | |||
| AK074612.1 | ||||
| AK223566.1 | BAD97286.1 | |||
| AK301637.1 | ||||
| AK303148.1 | ||||
| AK312731.1 | ||||
| AY102608.1 | AAM28950.1 | |||
| BC005143.2 | AAH05143.1 | |||
| BC020797.1 | AAH20797.1 |
More Information
Set Membership: |
|

Genomic Map