Assay Details
Target Gene Details
Entrez Gene ID: | 4435 |
Gene Name: | Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 1 |
Gene Aliases: |
MSG1 |
Location: |
Chr.X:72301638-72307187 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CITED1 | NM_001144885.1 | 4 | 837 | NP_001138357.1 |
| NM_001144886.1 | 3 | 789 | NP_001138358.1 | |
| NM_001144887.1 | 3 | 716 | NP_001138359.1 | |
| NM_004143.3 | 3 | 1137 | NP_004134.2 | |
| XM_011530958.1 | 4 | 828 | XP_011529260.1 | |
| AB451286.1 | ||||
| AB451418.1 | ||||
| AI217516.1 | 1 | 115 | ||
| BC004240.1 | 3 | 1090 | AAH04240.1 | |
| U65092.1 | 3 | 756 | AAC51113.1 |
More Information
Set Membership: |
|

Genomic Map