Assay Details
Target Gene Details
Entrez Gene ID: | 3309 |
Gene Name: | heat shock protein family A (Hsp70) member 5 |
Gene Aliases: |
BIP, GRP78, HEL-S-89n, MIF2 |
Location: |
Chr.9:125234848-125241387 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPA5 | NM_005347.4 | NP_005338.1 | ||
| AF188611.1 | AAF13605.1 | |||
| AF216292.1 | AAF42836.1 | |||
| AJ271729.1 | CAB71335.1 | |||
| AK293604.1 | ||||
| BC020235.1 | AAH20235.1 | |||
| DA549454.1 | ||||
| EU794617.1 | ||||
| X87949.1 | CAA61201.1 |
More Information
Set Membership: |
|

Genomic Map