Assay Details
Target Gene Details
Entrez Gene ID: | 340075 |
Gene Name: | arylsulfatase family member I |
Gene Aliases: |
SPG66 |
Location: |
Chr.5:150296346-150302962 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARSI | NM_001012301.2 | 2 | 2241 | NP_001012301.1 |
| AB448735.1 | 2 | 2232 | ||
| AK122641.1 | 2 | 1385 | ||
| AY875937.1 | 2 | 1652 | AAW66665.1 | |
| BC129995.1 | 2 | 1709 | ||
| BC129996.1 | 2 | 1709 |
More Information
Set Membership: |
|

Genomic Map