Assay Details
Target Gene Details
Entrez Gene ID: | 51247 |
Gene Name: | poly(A) binding protein interacting protein 2 |
Gene Aliases: |
PAIP-2, PAIP2A |
Location: |
Chr.5:139341832-139369720 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAIP2 | NM_001033112.2 | 4 | 836 | NP_001028284.1 |
| NM_016480.4 | 4 | 802 | NP_057564.3 | |
| NR_109830.1 | 4 | 798 | ||
| XM_011543428.1 | 5 | 884 | XP_011541730.1 | |
| XM_017009522.1 | 4 | 2210 | XP_016865011.1 | |
| XM_017009523.1 | 4 | 755 | XP_016865012.1 | |
| AF151052.1 | 4 | 760 | AAF36138.1 | |
| AF317675.1 | 4 | 712 | AAK11562.1 | |
| AK024552.1 | 4 | 776 | ||
| AL136640.1 | 4 | 776 | CAB66575.1 | |
| BC039337.1 | 4 | 1550 | AAH39337.1 | |
| BC048106.1 | 4 | 1619 | AAH48106.1 | |
| BC062718.1 | 4 | 824 | AAH62718.1 | |
| BI552800.1 | 4 | 807 | ||
| BU732743.1 | 1 | 697 |
Target Gene Details
Entrez Gene ID: | 9963 |
Gene Name: | solute carrier family 23 member 1 |
Gene Aliases: |
SLC23A2, SVCT1, YSPL3 |
Location: |
Chr.5:139367196-139384553 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC23A1 | NM_005847.4 | NP_005838.3 | ||
| NM_152685.3 | NP_689898.2 | |||
| XM_005272148.3 | XP_005272205.3 | |||
| XM_005272149.3 | XP_005272206.3 | |||
| XM_006714741.2 | XP_006714804.2 | |||
| XM_011543766.1 | XP_011542068.1 | |||
| XM_011543767.1 | XP_011542069.1 | |||
| XM_011543768.1 | XP_011542070.1 | |||
| XM_011543769.1 | XP_011542071.1 | |||
| AF098277.1 | AAF22490.1 | |||
| AF170911.1 | AAF24759.1 | |||
| AK092655.1 | ||||
| AK290789.1 | ||||
| BC019225.1 | AAH19225.1 | |||
| BC050261.1 | AAH50261.1 |
More Information
Set Membership: |
|

Genomic Map