Assay Details
Target Gene Details
Entrez Gene ID: | 57180 |
Gene Name: | ARP3 actin related protein 3 homolog B |
Gene Aliases: |
ARP11, ARP3BETA |
Location: |
Chr.7:152759749-153070979 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTR3B | NM_001040135.2 | NP_001035225.1 | ||
| NM_020445.5 | NP_065178.1 | |||
| NR_073000.1 | ||||
| NR_073001.1 | ||||
| XM_006716065.2 | XP_006716128.1 | |||
| XM_006716066.1 | XP_006716129.1 | |||
| XM_006716067.3 | XP_006716130.1 | |||
| XM_011516425.2 | XP_011514727.1 | |||
| XM_011516426.2 | XP_011514728.1 | |||
| XM_011516427.1 | XP_011514729.1 | |||
| XM_017012460.1 | XP_016867949.1 | |||
| XM_017012461.1 | XP_016867950.1 | |||
| XM_017012462.1 | XP_016867951.1 | |||
| XM_017012464.1 | XP_016867953.1 | |||
| AF023453.1 | AAC98904.1 | |||
| AF086920.1 | AAP97150.1 | |||
| AK294292.1 | ||||
| AK315534.1 | ||||
| AK316078.1 | ||||
| BC008682.1 | AAH08682.1 | |||
| BC015207.1 | AAH15207.1 | |||
| DN601600.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3615528 | Chr.7:152741193 - 152762891 on Build GRCh38 | Gain |
|
| esv3615526 | Chr.7:152717766 - 152770123 on Build GRCh38 | Gain |
|
| nsv831193 | Chr.7:152656459 - 152895593 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map