Assay Details
Target Gene Details
Entrez Gene ID: | 85369 |
Gene Name: | striatin interacting protein 1 |
Gene Aliases: |
FAM40A, FAR11A |
Location: |
Chr.1:110031577-110054641 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STRIP1 | NM_001270768.1 | 9 | 892 | NP_001257697.1 |
| NM_033088.3 | 9 | 1045 | NP_149079.2 | |
| NR_073071.1 | 10 | 1159 | ||
| XM_006710995.2 | 2 | 148 | XP_006711058.1 | |
| XM_017002629.1 | 9 | 1044 | XP_016858118.1 | |
| XM_017002630.1 | 9 | 1044 | XP_016858119.1 | |
| AB051548.2 | 9 | 1010 | BAB21852.2 | |
| AK027649.1 | 9 | 892 | BAB55265.1 | |
| AK125054.1 | 2 | 814 | BAC86034.1 | |
| AK291059.1 | 9 | 1019 | ||
| AL834196.1 | 10 | 1128 | CAH56368.1 | |
| BC094786.1 | 9 | 1026 | AAH94786.1 | |
| BC119814.1 | 9 | 1027 | ||
| BC121793.1 | 9 | 1027 |
More Information
Set Membership: |
|

Genomic Map