Assay Details
Target Gene Details
Entrez Gene ID: | 260429 |
Gene Name: | protease, serine 33 |
Gene Aliases: |
EOS |
Location: |
Chr.16:2783953-2787759 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS33 | NM_152891.2 | 5 | 787 | NP_690851.2 |
| XM_011522451.2 | 6 | 911 | XP_011520753.1 | |
| XM_011522452.2 | 6 | 723 | XP_011520754.1 | |
| AF536382.1 | 5 | 695 | AAN04055.1 | |
| BC036846.1 | 6 | 769 | AAH36846.2 | |
| BC062334.1 | 5 | 789 | AAH62334.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482187 | Chr.16:2783953 - 2786707 on Build GRCh38 | Gain |
|
| esv2760067 | Chr.16:2486824 - 2820535 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map