Assay Details
Target Gene Details
Entrez Gene ID: | 51093 |
Gene Name: | ribosomal RNA adenine dimethylase domain containing 1 |
Gene Aliases: |
C1orf66, CGI-41, METTL25B |
Location: |
Chr.1:156728471-156736960 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRNAD1 | NM_001142560.1 | NP_001136032.1 | ||
| NM_015997.3 | NP_057081.3 | |||
| XM_005245234.4 | XP_005245291.1 | |||
| XM_005245235.4 | XP_005245292.1 | |||
| XM_005245236.4 | XP_005245293.1 | |||
| XM_005245237.4 | XP_005245294.1 | |||
| XM_011509600.1 | XP_011507902.1 | |||
| XM_017001404.1 | XP_016856893.1 | |||
| XM_017001405.1 | XP_016856894.1 | |||
| XM_017001406.1 | XP_016856895.1 | |||
| AF151800.1 | AAD34037.1 | |||
| AK000412.1 | ||||
| BC011382.1 | AAH11382.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831648 | Chr.1:156694085 - 156912126 on Build GRCh38 | Loss |
|
| esv3578200 | Chr.1:156731053 - 156740837 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map