Assay Details
Target Gene Details
Entrez Gene ID: | 596 |
Gene Name: | BCL2, apoptosis regulator |
Gene Aliases: |
Bcl-2, PPP1R50 |
Location: |
Chr.18:63123346-63319778 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL2 | NM_000633.2 | 3 | 2102 | NP_000624.2 |
| BC027258.1 | 3 | 1998 | AAH27258.1 | |
| M13994.1 | 2 | 3067 | AAA51813.1 | |
| M14745.1 | 2 | 1640 | AAA35591.1 |
More Information
Set Membership: |
|

Genomic Map