Assay Details
Target Gene Details
Entrez Gene ID: | 28991 |
Gene Name: | COMM domain containing 5 |
Gene Aliases: |
HCARG, HT002 |
Location: |
Chr.8:144837978-144853578 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COMMD5 | AK023070.1 | 3 | 1995 | BAB14389.1 |
Target Gene Details
Entrez Gene ID: | 7553 |
Gene Name: | zinc finger protein 7 |
Gene Aliases: |
HF.16, KOX4, zf30 |
Location: |
Chr.8:144827500-144848070 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023752 | Chr.8:144835600 - 144866312 on Build GRCh38 | Gain |
|
| nsv522459 | Chr.8:144519234 - 144845293 on Build GRCh38 | Loss |
|
| nsv1031737 | Chr.8:144830982 - 144986209 on Build GRCh38 | Gain |
|
| nsv612969 | Chr.8:144785726 - 144877525 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map