Assay Details
Target Gene Details
Entrez Gene ID: | 51171 |
Gene Name: | hydroxysteroid 17-beta dehydrogenase 14 |
Gene Aliases: |
DHRS10, SDR47C1, retSDR3 |
Location: |
Chr.19:48813014-48836688 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSD17B14 | NM_016246.2 | NP_057330.2 | ||
| XM_005258969.3 | XP_005259026.1 | |||
| XM_005258970.3 | XP_005259027.1 | |||
| AF126781.1 | AAF06940.1 | |||
| AK307437.1 | ||||
| AY358430.1 | AAQ88796.1 | |||
| BC006283.2 | AAH06283.1 | |||
| BC006294.1 | AAH06294.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| nsv833855 | Chr.19:48750098 - 48949781 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map