Assay Details
Target Gene Details
Entrez Gene ID: | 57678 |
Gene Name: | glycerol-3-phosphate acyltransferase, mitochondrial |
Gene Aliases: |
GPAT, GPAT1 |
Location: |
Chr.10:112149864-112215395 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPAM | NM_001244949.1 | 22 | 3401 | NP_001231878.1 |
| NM_020918.5 | 22 | 3397 | NP_065969.3 | |
| XM_005269998.1 | 22 | 3542 | XP_005270055.1 | |
| AB046780.1 | 16 | 2689 | BAB13386.1 | |
| AK172782.1 | 10 | 1954 | BAD18763.1 | |
| AL833061.2 | 22 | 3383 | CAD89967.1 | |
| AL833093.2 | 22 | 3386 | CAD89932.1 | |
| BC030783.1 | 22 | 3365 | AAH30783.1 | |
| BX647451.1 | 22 | 3365 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508608 | Chr.10:112030892 - 112158412 on Build GRCh38 | Deletion |
|
| nsv831989 | Chr.10:112112194 - 112301601 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map