Assay Details
Target Gene Details
Entrez Gene ID: | 832 |
Gene Name: | capping actin protein of muscle Z-line beta subunit |
Gene Aliases: |
CAPB, CAPPB, CAPZ |
Location: |
Chr.1:19338773-19485641 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPZB | NM_001206540.2 | NP_001193469.1 | ||
| NM_001206541.2 | NP_001193470.1 | |||
| NM_001282162.1 | NP_001269091.1 | |||
| NM_004930.4 | NP_004921.1 | |||
| XM_006710938.3 | XP_006711001.1 | |||
| XM_011542228.2 | XP_011540530.1 | |||
| XM_011542229.2 | XP_011540531.1 | |||
| XM_011542230.2 | XP_011540532.1 | |||
| XM_017002428.1 | XP_016857917.1 | |||
| AK125085.1 | ||||
| AK301442.1 | ||||
| AK313113.1 | ||||
| BC024601.1 | AAH24601.1 | |||
| BC107752.1 | AAI07753.1 | |||
| BC109241.1 | AAI09242.1 | |||
| BC109242.1 | AAI09243.1 | |||
| BE870234.1 | ||||
| BT019470.1 | AAV38277.1 | |||
| BT019471.1 | AAV38278.1 | |||
| CB267021.1 | ||||
| EB387955.1 | ||||
| U03271.1 | AAA87395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv21e212 | Chr.1:19324106 - 19361938 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map