Assay Details
Target Gene Details
Entrez Gene ID: | 7071 |
Gene Name: | Kruppel like factor 10 |
Gene Aliases: |
EGR-alpha, EGRA, TIEG, TIEG1 |
Location: |
Chr.8:102648777-102655902 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLF10 | NM_005655.3 | NP_005646.1 | ||
| AK312894.1 | ||||
| BC095399.1 | AAH95399.1 | |||
| BI560615.1 | ||||
| DA720479.1 | ||||
| U21847.1 | AAC50340.1 |
Target Gene Details
Entrez Gene ID: | 101927245 |
Gene Name: | vegetative cell wall protein gp1-like |
Gene Aliases: |
- |
Location: |
Chr.8:102654313-102730620 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927245 | XM_011517419.2 | XP_011515721.1 | ||
| XM_011517420.2 | XP_011515722.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33118 | Chr.8:102346339 - 103055944 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map