Assay Details
Target Gene Details
Entrez Gene ID: | 6276 |
Gene Name: | S100 calcium binding protein A5 |
Gene Aliases: |
S100D |
Location: |
Chr.1:153537114-153543514 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A5 | NM_002962.1 | 3 | 304 | NP_002953.2 |
| XM_011509860.2 | 2 | 794 | XP_011508162.1 | |
| XM_017002029.1 | 3 | 398 | XP_016857518.1 | |
| XM_017002030.1 | 3 | 727 | XP_016857519.1 | |
| XM_017002031.1 | 2 | 1040 | XP_016857520.1 | |
| XM_017002032.1 | 2 | 923 | XP_016857521.1 | |
| BC093955.1 | 3 | 160 | AAH93955.1 | |
| BC093957.2 | 3 | 160 | AAH93957.1 | |
| Z18954.1 | 3 | 304 | CAA79479.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831603 | Chr.1:153433366 - 153603667 on Build GRCh38 | Gain |
|
| nsv1002705 | Chr.1:153335376 - 153553113 on Build GRCh38 | Gain |
|
| esv3587591 | Chr.1:153528630 - 153570193 on Build GRCh38 | Gain |
|
| esv3587584 | Chr.1:153466126 - 153598991 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map