Assay Details
Target Gene Details
Entrez Gene ID: | 84985 |
Gene Name: | family with sequence similarity 83 member A |
Gene Aliases: |
BJ-TSA-9 |
Location: |
Chr.8:123179047-123210079 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM83A | NM_001288587.2 | NP_001275516.1 | ||
| NM_001321630.1 | NP_001308559.1 | |||
| NM_032899.6 | NP_116288.2 | |||
| NM_207006.2 | NP_996889.1 | |||
| XM_005251087.2 | XP_005251144.1 | |||
| XM_011517338.1 | XP_011515640.1 | |||
| AF497803.1 | AAQ07260.1 | |||
| AK095301.1 | ||||
| AK098407.1 | BAC05299.1 | |||
| BC007828.2 | AAH07828.1 | |||
| BC052300.1 | AAH52300.1 | |||
| DQ280322.1 | ||||
| DQ280323.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1032827 | Chr.8:122655842 - 123258160 on Build GRCh38 | Gain |
|
| esv3618663 | Chr.8:123180821 - 123192499 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map