Assay Details
Target Gene Details
Entrez Gene ID: | 6340 |
Gene Name: | sodium channel epithelial 1 gamma subunit |
Gene Aliases: |
BESC3, ENaCg, ENaCgamma, PHA1, SCNEG |
Location: |
Chr.16:23182719-23216879 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCNN1G | NM_001039.3 | 2 | 225 | NP_001030.2 |
| BC059391.1 | 2 | 225 | AAH59391.1 | |
| BC069652.1 | 1 | 86 | AAH69652.1 | |
| DQ402524.1 | 1 | 86 | ||
| L36592.1 | 1 | 108 | AAA75460.1 | |
| U48936.1 | 2 | 227 | AAC50737.1 | |
| X87160.1 | 1 | 86 | CAA60633.1 |
More Information
Set Membership: |
|

Genomic Map