Assay Details
Target Gene Details
Entrez Gene ID: | 84915 |
Gene Name: | family with sequence similarity 222 member A |
Gene Aliases: |
C12orf34 |
Location: |
Chr.12:109714382-109770509 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM222A | NM_032829.2 | 3 | 743 | NP_116218.2 |
| XM_006719654.2 | 3 | 329 | XP_006719717.1 | |
| XM_017020055.1 | 3 | 713 | XP_016875544.1 | |
| AK027627.1 | 3 | 347 | BAB55244.1 | |
| AK074806.1 | BAC11219.1 | |||
| BC037221.1 | 2 | 218 | AAH37221.1 |
Target Gene Details
Entrez Gene ID: | 84983 |
Gene Name: | FAM222A antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:109734209-109773487 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM222A-AS1 | NR_026661.2 | |||
| NR_026662.2 | ||||
| BQ684893.1 | ||||
| DA082338.1 |
More Information
Set Membership: |
|

Genomic Map