Assay Details
Target Gene Details
Entrez Gene ID: | 51340 |
Gene Name: | crooked neck pre-mRNA splicing factor 1 |
Gene Aliases: |
CLF, CRN, Clf1, HCRN, MSTP021, SYF3 |
Location: |
Chr.20:20034361-20056046 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRNKL1 | NM_001278625.1 | 12 | 1871 | NP_001265554.1 |
| NM_001278626.1 | 12 | 1684 | NP_001265555.1 | |
| NM_001278627.1 | 12 | 1594 | NP_001265556.1 | |
| NM_001278628.1 | 11 | 1542 | NP_001265557.1 | |
| NM_016652.5 | 12 | 1907 | NP_057736.4 | |
| AB016840.1 | 11 | 1504 | BAC53587.1 | |
| AF111802.1 | 11 | 1498 | AAL39004.1 | |
| AF255443.2 | 12 | 1871 | AAF65571.2 | |
| AF318302.1 | 11 | 1489 | AAK01924.1 | |
| AF318303.1 | 12 | 1907 | AAK01925.1 | |
| AF318304.1 | 12 | 1684 | AAK01926.1 | |
| AF318305.1 | 12 | 1594 | AAK01927.1 | |
| AK022908.1 | 10 | 1219 | BAB14303.1 | |
| AK023246.1 | 8 | 1054 | BAB14485.1 | |
| AK023728.1 | 7 | 868 | BAB14659.1 | |
| AK292799.1 | 11 | 1412 | ||
| AL832435.1 | 11 | 1516 | CAH10656.1 |
More Information
Set Membership: |
|

Genomic Map