Assay Details
Target Gene Details
Entrez Gene ID: | 23483 |
Gene Name: | TDP-glucose 4,6-dehydratase |
Gene Aliases: |
CATMANS, SDR2E1, TDPGD |
Location: |
Chr.13:94574054-94596275 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGDS | NM_001304430.1 | NP_001291359.1 | ||
| NM_014305.3 | NP_055120.1 | |||
| NR_130731.1 | ||||
| NR_130732.1 | ||||
| XM_011521065.2 | XP_011519367.1 | |||
| XM_011521066.2 | XP_011519368.1 | |||
| AF048686.1 | AAD50061.1 | |||
| AJ006068.1 | CAA06840.1 | |||
| AK303439.1 | ||||
| AK313251.1 | ||||
| AL527626.3 | ||||
| AY544125.1 | AAT11156.1 | |||
| BC005284.1 | ||||
| BC033675.1 | AAH33675.1 | |||
| BG546428.1 | ||||
| BG702561.1 | ||||
| BI914642.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832686 | Chr.13:94554488 - 94748738 on Build GRCh38 | Gain |
|
| nsv832685 | Chr.13:94527422 - 94627712 on Build GRCh38 | Gain |
|
| esv34004 | Chr.13:94254283 - 94740390 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map