Assay Details
Target Gene Details
Entrez Gene ID: | 102465248 |
Gene Name: | microRNA 6501 |
Gene Aliases: |
hsa-mir-6501, mir-6501 |
Location: |
Chr.21:33550662-33550728 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR6501 | NR_106756.1 |
Target Gene Details
Entrez Gene ID: | 6651 |
Gene Name: | SON DNA binding protein |
Gene Aliases: |
BASS1, C21orf50, DBP-5, NREBP, SON3 |
Location: |
Chr.21:33543038-33577514 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SON | NM_001291411.1 | 3 | 1507 | NP_001278340.1 |
| NM_001291412.1 | NP_001278341.1 | |||
| NM_032195.2 | 3 | 1507 | NP_115571.2 | |
| NM_138927.2 | 3 | 1507 | NP_620305.2 | |
| NR_103797.1 | 3 | 1507 | ||
| AB028942.2 | 3 | 1476 | ||
| AF380179.1 | 3 | 1481 | AAL34497.1 | |
| AF380180.1 | 3 | 1481 | AAL34498.1 | |
| AF380181.1 | 3 | 1481 | AAL34499.1 | |
| AF380183.1 | 3 | 1481 | AAL34501.1 | |
| AF380184.1 | 3 | 1481 | AAL34502.1 | |
| AF435977.1 | 3 | 1467 | AAL30810.1 | |
| AK074269.1 | 1 | 468 | ||
| AK091233.1 | ||||
| AK127947.1 | BAC87202.1 | |||
| AK128071.1 | 1 | 454 | ||
| AK307612.1 | 3 | 1507 | ||
| AY026895.1 | 3 | 1501 | AAK07692.1 | |
| BC002422.2 | AAH02422.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1115615 | Chr.21:33548994 - 33557394 on Build GRCh38 | Duplication |
|
| nsv1078502 | Chr.21:33549893 - 33553894 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map