Assay Details
Target Gene Details
Entrez Gene ID: | 3241 |
Gene Name: | hippocalcin like 1 |
Gene Aliases: |
BDR1, HLP2, VILIP-3 |
Location: |
Chr.2:10302124-10427617 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPCAL1 | NM_002149.3 | 1 | 102 | NP_002140.2 |
| XM_005246160.1 | 1 | 117 | XP_005246217.1 | |
| XM_005246161.1 | 1 | 117 | XP_005246218.1 | |
| XM_005246162.1 | 1 | 117 | XP_005246219.1 | |
| XM_005246163.1 | 1 | 117 | XP_005246220.1 | |
| XM_011510347.1 | 1 | 117 | XP_011508649.1 | |
| XM_011510349.1 | 1 | 117 | XP_011508651.1 | |
| XM_017003950.1 | 1 | 117 | XP_016859439.1 | |
| XM_017003951.1 | 1 | 117 | XP_016859440.1 | |
| XM_017003952.1 | XP_016859441.1 | |||
| BC009846.2 | 1 | 77 | AAH09846.1 | |
| BC017028.1 | 1 | 69 | AAH17028.1 | |
| BC017482.1 | 1 | 92 | AAH17482.1 | |
| BI603158.1 | 1 | 107 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv27364 | Chr.2:10302345 - 10303673 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map