Assay Details
Target Gene Details
Entrez Gene ID: | 57835 |
Gene Name: | solute carrier family 4 member 5 |
Gene Aliases: |
NBC4, NBCe2 |
Location: |
Chr.2:74216242-74343407 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC4A5 | NM_021196.3 | 19 | 2697 | NP_067019.3 |
| NM_133478.2 | 24 | 3093 | NP_597812.1 | |
| AB209752.1 | BAD92989.1 | |||
| AF243499.2 | 19 | 2695 | AAK26741.1 | |
| AF293337.2 | 23 | 3042 | AAK97072.1 | |
| AF293338.2 | 19 | 2695 | AAK97073.1 | |
| AF452248.1 | 21 | 2876 | AAL48291.1 | |
| AF453528.1 | AAL50802.1 | |||
| BC109221.2 | 18 | 2438 | AAI09222.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834261 | Chr.2:74164582 - 74386593 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map