Assay Details
Target Gene Details
Entrez Gene ID: | 84618 |
Gene Name: | 5'-nucleotidase, cytosolic IA |
Gene Aliases: |
CN-I, CN-IA, CN1, CN1A, CNI |
Location: |
Chr.1:39659121-39672038 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NT5C1A | NM_032526.2 | NP_115915.1 | ||
| AF331801.1 | AAK01294.1 | |||
| AY028778.1 | AAK30000.1 | |||
| BC103879.1 | AAI03880.1 | |||
| BC103880.1 | AAI03881.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv577 | Chr.1:39644349 - 39676840 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map